siRNA Details
| Gene Symbol: Gm5800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034102.2 | NP_001029274.1 | 3, 4 | 314 | ||
| GenBank mRNA | BC049651.1 | - | 3, 4 | 314 | ||
| BC100479.1 | - | 3, 4 | 314 | 30917737 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5800| Gene Symbol: Gm5800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034102.2 | NP_001029274.1 | 3, 4 | 314 | ||
| GenBank mRNA | BC049651.1 | - | 3, 4 | 314 | ||
| BC100479.1 | - | 3, 4 | 314 | 30917737 | ||