siRNA Details
| Gene Symbol: Olfr110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146328.2 | NP_666440.2 | 2 | 484 | ||
| GenBank mRNA | BC132347.1 | - | 1 | 428 | 40130624 | |
| Gene Symbol: Olfr111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005485.2 | NP_001005485.1 | 1 | 422 | ||
| GenBank mRNA | BC139116.1 | - | 1 | 422 | 8862138 | |
| BC139117.1 | - | 1 | 422 | 8862139 | ||

Olfr110
Genomic Map