siRNA Details
| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024717.2 | NP_001019888.1 | 3 | 598 | ||
| GenBank mRNA | BC066996.1 | - | 3 | 588 | 5707267 | |
| BU612752.1 | - | 3 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal3st3| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024717.2 | NP_001019888.1 | 3 | 598 | ||
| GenBank mRNA | BC066996.1 | - | 3 | 588 | 5707267 | |
| BU612752.1 | - | 3 | 598 | |||