siRNA Details
| Gene Symbol: Cyp2c67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024719.2 | NP_001019890.1 | 2 | 235 | ||
| GenBank mRNA | AK149389.1 | - | 2 | 246 | ||
| BC092260.1 | - | 2 | 235 | 5132591 | ||
| BC106142.1 | - | 2 | 235 | 5051946 | ||

Cyp2c67
Genomic Map