siRNA Details
Gene Symbol: Sh2d1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033499.1 | NP_001028671.1 | 3 | 337 | ||
GenBank mRNA | DQ131181.1 | - | 3 | 337 | ||
DQ140178.1 | - | 3 | 337 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sh2d1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033499.1 | NP_001028671.1 | 3 | 337 | ||
GenBank mRNA | DQ131181.1 | - | 3 | 337 | ||
DQ140178.1 | - | 3 | 337 |