siRNA Details
| Gene Symbol: B020018G12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139805.1 | - | 1 | 383 | ||
| BC132651.1 | - | 1 | 372 | 40130928 | ||
| BC132653.1 | - | 1 | 372 | 40130930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B020018G12Rik| Gene Symbol: B020018G12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139805.1 | - | 1 | 383 | ||
| BC132651.1 | - | 1 | 372 | 40130928 | ||
| BC132653.1 | - | 1 | 372 | 40130930 | ||