siRNA Details
| Gene Symbol: Olfr1065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146408.2 | NP_666520.2 | 1 | 417 | ||
| GenBank mRNA | AF178750.1 | - | 1 | 39 | ||
| BC138401.1 | - | 1 | 417 | 8861421 | ||
| BC138402.1 | - | 1 | 417 | 8861422 | ||
| Gene Symbol: Olfr1058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146391.2 | NP_666503.2 | 1 | 393 | ||

Olfr1065
Genomic Map