siRNA Details
| Gene Symbol: Zfp345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034900.3 | NP_001030072.2 | 4 | 1547 | ||
| GenBank mRNA | AK144819.1 | - | 4 | 1548 | ||
| BC150850.1 | - | 4 | 1536 | 9087761 | ||
| BC150852.1 | - | 4 | 1536 | 9087763 | ||

Zfp345
Genomic Map