siRNA Details
| Gene Symbol: Gm12429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277167.1 | NP_001264096.1 | 4 | 3876 | ||
| GenBank mRNA | AV257990.2 | - | 1 | 417 | ||
| Gene Symbol: Gm13298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085530.1 | NP_001078999.1 | 4 | 3858 | ||

Gm12429
Genomic Map