siRNA Details
| Gene Symbol: B020004J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033790.3 | NP_001028962.1 | 3 | 707 | ||
| GenBank mRNA | BC147376.1 | - | 2 | 603 | 9056473 | |
| BC147377.1 | - | 2 | 603 | 9056471 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B020004J07Rik| Gene Symbol: B020004J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033790.3 | NP_001028962.1 | 3 | 707 | ||
| GenBank mRNA | BC147376.1 | - | 2 | 603 | 9056473 | |
| BC147377.1 | - | 2 | 603 | 9056471 | ||