siRNA Details
| Gene Symbol: Cyp4a31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252539.1 | NP_001239468.1 | 5 | 725 | ||
| NM_201640.3 | NP_964002.2 | 6 | 770 | |||
| GenBank mRNA | BC013476.1 | - | 5 | 676 | 4237837 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4a31| Gene Symbol: Cyp4a31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252539.1 | NP_001239468.1 | 5 | 725 | ||
| NM_201640.3 | NP_964002.2 | 6 | 770 | |||
| GenBank mRNA | BC013476.1 | - | 5 | 676 | 4237837 | |