siRNA Details
| Gene Symbol: Cyp4a31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252539.1 | NP_001239468.1 | 6 | 825 | ||
| NM_201640.3 | NP_964002.2 | 7 | 870 | |||
| GenBank mRNA | BC013476.1 | - | 6 | 776 | 4237837 | |
| Gene Symbol: Cyp4a32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100181.1 | NP_001093651.1 | 7 | 839 | ||

Cyp4a31
Genomic Map