siRNA Details
| Gene Symbol: RNASE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282192.1 | NP_001269121.1 | 3 | 851 | ||
| NM_001282193.1 | NP_001269122.1 | 2 | 685 | |||
| NM_002937.4 | NP_002928.1 | 2 | 747 | |||
| NM_194431.2 | NP_919412.1 | 2 | 299 | |||
| GenBank mRNA | AK092422.1 | - | 2 | 747 | ||
| BC015520.1 | - | 2 | 356 | 3905439 | ||
| BG616902.1 | - | 2 | 296 | |||
| BU594423.1 | - | 2 | 318 | |||
| BX247985.1 | - | 2 | 537 | |||
| BX248010.1 | - | 3 | 405 | |||
| D37931.1 | - | 1 | 211 | |||
| DA925725.1 | - | 3 | 477 | |||
| DR003809.1 | - | 2 | 302 | |||

RNASE4
Genomic Map