siRNA Details
Gene Symbol: Gm12888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033791.3 | NP_001028963.1 | 3 | 340 | ||
GenBank mRNA | AK136549.1 | - | 3 | 341 | ||
BC150899.1 | - | 3 | 341 | 9087810 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm12888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033791.3 | NP_001028963.1 | 3 | 340 | ||
GenBank mRNA | AK136549.1 | - | 3 | 341 | ||
BC150899.1 | - | 3 | 341 | 9087810 |