siRNA Details
| Gene Symbol: Gm5868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024147.2 | NP_001019318.1 | 3 | 1148 | ||
| GenBank mRNA | AK084047.1 | - | 3 | 1149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5868| Gene Symbol: Gm5868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024147.2 | NP_001019318.1 | 3 | 1148 | ||
| GenBank mRNA | AK084047.1 | - | 3 | 1149 | ||