siRNA Details
| Gene Symbol: Sec61b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024171.2 | NP_077133.1 | 4 | 452 | ||
| GenBank mRNA | AK004505.1 | - | 4 | 453 | ||
| AK008468.2 | - | 895 | ||||
| BC002089.1 | - | 4 | 424 | 3487410 | ||
| BC081445.1 | - | 4 | 420 | 6412192 | ||

Sec61b
Genomic Map