siRNA Details
| Gene Symbol: Igk-V8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF043114.1 | - | 149 | |||
| AF043116.1 | - | 149 | ||||
| U28969.1 | - | 149 | ||||
| Gene Symbol: Igkv14-111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF139221.1 | - | 1 | 137 | ||
| X70263.1 | - | 1 | 139 | |||

Igk-V8
Genomic Map