siRNA Details
| Gene Symbol: Gm5885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185040.1 | NP_001171969.1 | 3 | 512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5885| Gene Symbol: Gm5885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185040.1 | NP_001171969.1 | 3 | 512 | ||