siRNA Details
| Gene Symbol: Zscan4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100186.1 | NP_001093656.1 | 4 | 1305 | ||
| GenBank mRNA | EF143406.1 | - | 4 | 1305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zscan4d| Gene Symbol: Zscan4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100186.1 | NP_001093656.1 | 4 | 1305 | ||
| GenBank mRNA | EF143406.1 | - | 4 | 1305 | ||