siRNA Details
| Gene Symbol: Psg27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037168.1 | NP_001032245.1 | 4 | 1132 | ||
| GenBank mRNA | BC100351.1 | - | 4 | 1132 | 30713102 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psg27| Gene Symbol: Psg27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037168.1 | NP_001032245.1 | 4 | 1132 | ||
| GenBank mRNA | BC100351.1 | - | 4 | 1132 | 30713102 | |