siRNA Details
| Gene Symbol: Olfr612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200027.1 | NP_001186956.1 | 1 | 339 | ||
| Gene Symbol: Olfr611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146727.2 | NP_666938.2 | 1 | 339 | ||

Olfr612
Genomic Map