siRNA Details
| Gene Symbol: BB114814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI662013.1 | - | 3 | 404 | ||
| DV051111.1 | - | 2 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB114814| Gene Symbol: BB114814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI662013.1 | - | 3 | 404 | ||
| DV051111.1 | - | 2 | 114 | |||