siRNA Details
| Gene Symbol: BB114814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI662013.1 | - | 5 | 766 | ||
| DV051111.1 | - | 4 | 475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BB114814| Gene Symbol: BB114814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BI662013.1 | - | 5 | 766 | ||
| DV051111.1 | - | 4 | 475 | |||