siRNA Details
| Gene Symbol: Spag11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034905.2 | NP_001030077.1 | 2 | 1356 | ||
| NM_001039563.3 | NP_001034652.2 | 2, 3 | 258 | |||
| NM_001286493.1 | NP_001273422.1 | 2, 3 | 261 | |||
| GenBank mRNA | AK136627.1 | - | 2 | 1357 | ||
| AY552530.1 | - | 2, 3 | 224 | |||
| BC145547.1 | - | 2, 3 | 258 | 9054111 | ||
| BC145549.1 | - | 2, 3 | 255 | 9054113 | ||
| BC145818.1 | - | 2, 3 | 255 | 40131121 | ||
| BC145820.1 | - | 2, 3 | 255 | 40131123 | ||
| DQ012055.1 | - | 2, 3 | 219 | |||

Spag11b
Genomic Map