siRNA Details
| Gene Symbol: Ubtfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033793.3 | NP_001028965.1 | 3 | 1126 | ||
| GenBank mRNA | AK139937.1 | - | 3 | 1127 | ||
| AK163257.1 | - | 2 | 1243 | |||
| BC139141.1 | - | 2 | 941 | 8862163 | ||
| BC145822.1 | - | 2 | 941 | 40131125 | ||

Ubtfl1
Genomic Map