siRNA Details
| Gene Symbol: 9230110F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029863.1 | NP_084139.1 | 2 | 229 | ||
| GenBank mRNA | AK020329.1 | - | 2 | 230 | ||
| Gene Symbol: Gm5916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167587.1 | NP_001161059.1 | 2 | 229 | ||

9230110F15Rik
Genomic Map