siRNA Details
| Gene Symbol: 7420426K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033983.3 | NP_001029155.1 | 1 | 351 | ||
| GenBank mRNA | AK135855.1 | - | 1 | 352 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

7420426K07Rik| Gene Symbol: 7420426K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033983.3 | NP_001029155.1 | 1 | 351 | ||
| GenBank mRNA | AK135855.1 | - | 1 | 352 | ||