siRNA Details
| Gene Symbol: Fbxw18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033794.3 | NP_001028966.1 | 4 | 418 | ||
| GenBank mRNA | AK136230.1 | - | 4 | 422 | ||
| AK162167.1 | - | 4 | 403 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw18| Gene Symbol: Fbxw18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033794.3 | NP_001028966.1 | 4 | 418 | ||
| GenBank mRNA | AK136230.1 | - | 4 | 422 | ||
| AK162167.1 | - | 4 | 403 | |||