siRNA Details
| Gene Symbol: Igkv4-68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF019943.1 | - | 140 | |||
| AY135151.1 | - | 140 | ||||
| DQ372799.1 | - | 197 | ||||
| L21017.1 | - | 1 | 140 | |||
| L21019.1 | - | 1 | 140 | |||
| Z18670.1 | - | 1 | 140 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AY186203.1 | - | 140 | Chr. - - [-] on Build | ||

Igkv4-68
Genomic Map