siRNA Details
| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080560.3 | NP_542127.1 | 2 | 119 | ||
| GenBank mRNA | AK005302.1 | - | 2 | 137 | ||
| AK161968.1 | - | 2 | 137 | |||
| AY039837.1 | - | 2 | 128 | |||
| BC034898.1 | - | 2 | 96 | 1382079 | ||
| BC052203.1 | - | 2 | 96 | |||
| BC067069.1 | - | 2 | 119 | 30550024 | ||
| BC132630.1 | - | 2 | 94 | 40130907 | ||
| BC138211.1 | - | 2 | 94 | 8861230 | ||
| Y09873.1 | - | 2 | 106 | |||

Ube2n
Genomic Map