siRNA Details
| Gene Symbol: Klhl33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166651.1 | NP_001160123.1 | 2 | 952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl33| Gene Symbol: Klhl33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166651.1 | NP_001160123.1 | 2 | 952 | ||