siRNA Details
| Gene Symbol: Gm20939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024731.2 | NP_001019902.2 | 5 | 943 | ||
| GenBank mRNA | BC070434.1 | - | 5 | 943 | 6851340 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20939| Gene Symbol: Gm20939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024731.2 | NP_001019902.2 | 5 | 943 | ||
| GenBank mRNA | BC070434.1 | - | 5 | 943 | 6851340 | |