siRNA Details
| Gene Symbol: Tmem181b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082120.1 | - | 1 | 1212 | ||
| BC032285.1 | - | 3 | 1422 | |||
| BC096660.1 | - | 3 | 1331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem181b-ps| Gene Symbol: Tmem181b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082120.1 | - | 1 | 1212 | ||
| BC032285.1 | - | 3 | 1422 | |||
| BC096660.1 | - | 3 | 1331 | |||