siRNA Details
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082120.1 | - | 1 | 932 | ||
BC032285.1 | - | 3 | 1145 | |||
BC096660.1 | - | 3 | 1054 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082120.1 | - | 1 | 932 | ||
BC032285.1 | - | 3 | 1145 | |||
BC096660.1 | - | 3 | 1054 |