siRNA Details
| Gene Symbol: ROM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000327.3 | NP_000318.1 | 1 | 1065 | ||
| GenBank mRNA | AK313674.1 | - | 1 | 1065 | ||
| BC008100.1 | - | 1 | 655 | 4177529 | ||
| BC036268.1 | - | 1 | 655 | |||
| L07894.1 | - | 1 | 529 | |||

ROM1
Genomic Map