siRNA Details
| Gene Symbol: Dnah8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013811.3 | NP_038839.2 | 5098 | |||
| GenBank mRNA | AF356520.1 | - | 5098 | |||
| AF356521.1 | - | 5098 | ||||
| AF356522.1 | - | 5108 | ||||
| AF356523.1 | - | 5108 | ||||
| AF363577.1 | - | 5108 | ||||

Dnah8
Genomic Map