siRNA Details
| Gene Symbol: Arid3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017362.2 | NP_001017362.1 | 5 | 704 | ||
| NM_001252622.1 | NP_001239551.1 | 5 | 704 | |||
| GenBank mRNA | BC141237.1 | - | 4 | 673 | 9055849 | |
| BC145563.1 | - | 4 | 673 | 9054128 | ||

Arid3c
Genomic Map