siRNA Details
| Gene Symbol: Xlr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081643.1 | NP_001075112.1 | 2 | 154 | ||
| GenBank mRNA | AK146398.1 | - | 2 | 123 | ||
| BC038862.1 | - | 2 | 117 | |||
| BC057003.1 | - | 2 | 126 | |||
| BC064787.1 | - | 2 | 111 | 30046437 | ||
| CJ063846.1 | - | 2 | 155 | |||
| Gene Symbol: Xlr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110784.1 | NP_001104254.1 | 2 | 198 | ||
| GenBank mRNA | BC003812.1 | - | 2 | 144 | 3601144 | |
| BC093495.1 | - | 121 | 3593889 | |||
| BC107255.1 | - | 107 | 40052405 | |||
| BC107283.1 | - | 108 | 40052875 | |||
| BC127145.1 | - | 108 | 40052876 | |||
| BC127146.1 | - | 109 | 40052877 | |||
| BQ885410.1 | - | 2 | 138 | |||
| DV662513.1 | - | 2 | 146 | |||
| U02599.1 | - | 1 | 76 | |||
| Gene Symbol: Xlr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011727.2 | NP_035857.2 | 2 | 123 | ||
| GenBank mRNA | L22977.1 | - | 108 | |||
| U02600.1 | - | 2 | 123 | |||

Xlr3b
Genomic Map