siRNA Details
| Gene Symbol: Zfp872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033813.4 | NP_001028985.1 | 4 | 758 | ||
| GenBank mRNA | AK137156.1 | - | 4 | 760 | ||
| BC150971.1 | - | 4 | 751 | 9087884 | ||
| BC151175.1 | - | 4 | 751 | 9088064 | ||

Zfp872
Genomic Map