siRNA Details
| Gene Symbol: Defb42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034910.3 | NP_001030082.1 | 3 | 308 | ||
| GenBank mRNA | AK079042.1 | - | 3 | 309 | ||
| BC146022.1 | - | 2 | 164 | 40131325 | ||
| BC146024.1 | - | 2 | 164 | 40131327 | ||
| DQ012046.1 | - | 3 | 308 | |||

Defb42
Genomic Map