siRNA Details
Gene Symbol: Gm6086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039219.2 | NP_001034308.1 | 8 | 1863 | ||
GenBank mRNA | AK145447.1 | - | 8 | 1864 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm6086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039219.2 | NP_001034308.1 | 8 | 1863 | ||
GenBank mRNA | AK145447.1 | - | 8 | 1864 |