siRNA Details
| Gene Symbol: RP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203288.1 | NP_976033.1 | 5, 6 | 493 | ||
| XM_011515468.2 | XP_011513770.1 | 5, 6 | 969 | |||
| GenBank mRNA | BC025928.1 | - | 5, 6 | 493 | 4903338 | |
| Gene Symbol: RP9P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003500.1 | 5, 6 | 577 | |||
| GenBank mRNA | AF274938.1 | - | 4, 5 | 287 | ||
| BC050570.1 | - | 6, 7 | 682 | |||

RP9
Genomic Map