siRNA Details
| Gene Symbol: Catsperd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175350.3 | NP_780559.2 | 4 | 252 | ||
| GenBank mRNA | AK014981.1 | - | 4 | 253 | ||
| HQ441159.1 | - | 4 | 244 | |||
| HQ441160.1 | - | 3 | 144 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Catsperd| Gene Symbol: Catsperd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175350.3 | NP_780559.2 | 4 | 252 | ||
| GenBank mRNA | AK014981.1 | - | 4 | 253 | ||
| HQ441159.1 | - | 4 | 244 | |||
| HQ441160.1 | - | 3 | 144 | |||