siRNA Details
Gene Symbol: Igkv5-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M16162.1 | - | 2 | 205 |
Gene Symbol: Igkv5-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY208309.1 | - | 139 | |||
U55592.1 | - | 1 | 132 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U55629.1 | - | 127 | Chr. - - [-] on Build |
