siRNA Details
| Gene Symbol: Dcpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077633.1 | NP_001071101.1 | 4 | 410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcpp3| Gene Symbol: Dcpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077633.1 | NP_001071101.1 | 4 | 410 | ||