siRNA Details
Gene Symbol: Vmn1r101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166836.1 | NP_001160308.1 | 1 | 514 |
Gene Symbol: Gm16442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167148.1 | NP_001160620.1 | 1 | 514 |
Gene Symbol: Gm16451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167149.1 | NP_001160621.1 | 1 | 514 |
Gene Symbol: Vmn1r149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166840.1 | NP_001160312.1 | 1 | 514 |
Gene Symbol: Vmn1r158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166841.1 | NP_001160313.1 | 1 | 514 |
