siRNA Details
| Gene Symbol: Gm8653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167152.1 | NP_001160624.1 | 1 | 252 | ||
| Gene Symbol: Gm6164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167153.1 | NP_001160625.1 | 1 | 252 | ||
| Gene Symbol: Vmn1r117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166743.1 | NP_001160215.1 | 1 | 252 | ||

Gm8653
Genomic Map