siRNA Details
Gene Symbol: Sult2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101586.2 | NP_001095056.2 | 1 | 115 |
Gene Symbol: Sult2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101534.1 | NP_001095004.1 | 1 | 112 |
Gene Symbol: Sult2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184980.1 | NP_001171909.1 | 1 | 193 | ||
GenBank mRNA | AI315523.1 | - | 1 | 193 |
