siRNA Details
| Gene Symbol: Sult2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184980.1 | NP_001171909.1 | 2 | 337 | ||
| GenBank mRNA | AI315523.1 | - | 2 | 337 | ||
| Gene Symbol: Sult2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101534.1 | NP_001095004.1 | 2 | 256 | ||

Sult2a5
Genomic Map