siRNA Details
| Gene Symbol: Ttc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081228.1 | NP_001074697.1 | 1 | 1928 | ||
| GenBank mRNA | BC157934.1 | - | 1 | 1949 | 9088113 | |
| BC158130.1 | - | 1 | 2045 | 9007398 | ||
| BC172090.1 | - | 1 | 1949 | 9054376 | ||

Ttc30a2
Genomic Map